ENAH5802F49B Enamine

2,6-dimethoxy-3-methylbenzene-1-sulfonyl chloride

  • MDL number MFCD28614881
  • Cas Number : 1785309-08-1
  • Molecular Weight : 250.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,6-dimethoxy-3-methylbenzenesulfonyl chloride
Inchi Code 1S/C9H11ClO4S/c1-6-4-5-7(13-2)9(8(6)14-3)15(10,11)12/h4-5H,1-3H3
Inchi Key RBMDAYLTRGXOKE-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7356239

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7356239
Availability details for
Quantity
  • Availability
  • Add to Cart