ENAH5802F4A0 Enamine

4-bromo-1-methyl-1H-pyrazole-3-sulfonyl chloride

  • MDL number MFCD28638419
  • Cas Number : 147544-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7266990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7266990
Availability details for
Quantity
  • Availability
  • Add to Cart