ENAH5802F4A4 Enamine

2-chloro-4-acetamidobenzene-1-sulfonyl chloride

  • MDL number MFCD09049250
  • Cas Number : 1954-95-6
  • Molecular Weight : 268.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 138-140
Iupac Name 4-(acetylamino)-2-chlorobenzenesulfonyl chloride
Inchi Code 1S/C8H7Cl2NO3S/c1-5(12)11-6-2-3-8(7(9)4-6)15(10,13)14/h2-4H,1H3,(H,11,12)
Inchi Key MSSDFXARSXTRSY-UHFFFAOYSA-N

Description

Melting Point 138-140
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-270686

Safety Information

signalword Danger
pcodes P260,P264,P280,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P363,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-270686
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart