ENAH5802F4A9 Enamine

[4-(trifluoromethyl)bicyclo[2.2.1]heptan-1-yl]methanesulfonyl chloride

  • MDL number MFCD32066418
  • Cas Number : 2361635-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6499671

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6499671
Availability details for
Quantity
  • Availability
  • Add to Cart