ENAH5802F4AB Enamine

2,6-difluoro-4-(trifluoromethyl)benzene-1-sulfonyl chloride

  • Cas Number : 2375261-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7443713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7443713
Availability details for
Quantity
  • Availability
  • Add to Cart