ENAH5802F4AC Enamine

2-bromo-3,4-dimethylbenzene-1-sulfonyl chloride

  • MDL number MFCD32066351
  • Cas Number : 2361635-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7431092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7431092
Availability details for
Quantity
  • Availability
  • Add to Cart