ENAH5802F4AF Enamine

3-bromo-5-chlorothiophene-2-sulfonyl chloride

  • MDL number MFCD00052095
  • Cas Number : 175205-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6941141

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6941141
Availability details for
Quantity
  • Availability
  • Add to Cart