ENAH5802F4B3 Enamine

1-(4-chlorophenyl)-3,5-dimethyl-1H-pyrazole-4-sulfonyl chloride

  • MDL number MFCD11967174
  • Cas Number : 1156603-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-272272

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-272272
Availability details for
Quantity
  • Availability
  • Add to Cart