ENAH5802F4B5 Enamine

tert-butyl 3-[(chlorosulfonyl)methyl]-3-cyanopyrrolidine-1-carboxylate

  • MDL number MFCD32066485
  • Cas Number : 2361645-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432027
Availability details for
Quantity
  • Availability
  • Add to Cart