ENAH5802F4BA Enamine

tert-butyl 3-[(chlorosulfonyl)methyl]-3-cyanopiperidine-1-carboxylate

  • MDL number MFCD32066030
  • Cas Number : 2361634-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7432020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7432020
Availability details for
Quantity
  • Availability
  • Add to Cart