ENAH5802F4BB Enamine

3-bromo-1-fluoronaphthalene-2-sulfonyl chloride

  • MDL number MFCD32065914
  • Cas Number : 2361636-25-9
  • Molecular Weight : 323.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-1-fluoronaphthalene-2-sulfonyl chloride
Inchi Code 1S/C10H5BrClFO2S/c11-8-5-6-3-1-2-4-7(6)9(13)10(8)16(12,14)15/h1-5H
Inchi Key IZBZIGYDYNGQOY-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7430688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7430688
Availability details for
Quantity
  • Availability
  • Add to Cart