ENAH5802F4C0 Enamine

3-[2-bromo-5-(chlorosulfonyl)-4-methoxyphenyl]propanoic acid

  • Cas Number : 2344685-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7355359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7355359
Availability details for
Quantity
  • Availability
  • Add to Cart