ENAH5802F4C1 Enamine

3,5-dibromo-2,6-dimethylbenzene-1-sulfonyl chloride

  • MDL number MFCD32069211
  • Cas Number : 343333-84-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6754457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6754457
Availability details for
Quantity
  • Availability
  • Add to Cart