ENAH5802F4C2 Enamine

4-methoxy-6-(trifluoromethyl)-1,3,5-triazin-2-amine

  • MDL number MFCD01679229

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6735250

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6735250
Availability details for
Quantity
  • Availability
  • Add to Cart