ENAH5802F4C4 Enamine

1H-pyrazole-3-thiol hydrochloride

  • MDL number MFCD32065930
  • Cas Number : 2361636-39-5
  • Molecular Weight : 136.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1H-pyrazole-3-thiol hydrochloride
Inchi Code 1S/C3H4N2S.ClH/c6-3-1-2-4-5-3;/h1-2H,(H2,4,5,6);1H
Inchi Key BPKRKGJLFFVZQI-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7433585

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7433585
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart