ENAH5802F4C7 Enamine

2-(2-aminoethoxy)ethane-1-thiol hydrochloride

  • Cas Number : 137048-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7424822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7424822
Availability details for
Quantity
  • Availability
  • Add to Cart