ENAH5802F4C9 Enamine

{bicyclo[3.1.0]hexan-6-yl}methanethiol

  • MDL number MFCD32198344
  • Cas Number : 2375273-93-9
  • Molecular Weight : 128.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name bicyclo[3.1.0]hexan-6-ylmethanethiol

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7310610

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7310610
Availability details for
Quantity
  • Availability
  • Add to Cart