ENAH5802F4CB Enamine

5-(1,2,2,2-tetrafluoroethyl)pyridin-2-amine hydrochloride

  • Cas Number : 2375262-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7440095

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440095
Availability details for
Quantity
  • Availability
  • Add to Cart