ENAH5802F4D3 Enamine

ethyl 5-sulfanyl-1H-pyrazole-4-carboxylate

  • MDL number MFCD32066461
  • Molecular Weight : 172.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-mercapto-1H-pyrazole-4-carboxylate
Inchi Code 1S/C6H8N2O2S/c1-2-10-6(9)4-3-7-8-5(4)11/h3H,2H2,1H3,(H2,7,8,11)
Inchi Key XDMLUSNSDZIDLF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6736241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6736241
Availability details for
Quantity
  • Availability
  • Add to Cart