ENAH5802F4D8 Enamine

2-amino-[1,3]thiazolo[5,4-d]pyrimidine-7-thiol

  • MDL number MFCD32197730
  • Cas Number : 2375261-14-4
  • Molecular Weight : 184.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-aminothiazolo[5,4-d]pyrimidine-7-thiol

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7440361

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440361
Availability details for
Quantity
  • Availability
  • Add to Cart