ENAH5802F4DC Enamine

3-(4-methyl-5-sulfanyl-4H-1,2,4-triazol-3-yl)propanoic acid

  • MDL number MFCD30378618
  • Cas Number : 178237-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-2010722

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-2010722
Availability details for
Quantity
  • Availability
  • Add to Cart