ENAH5802F4DE Enamine

4-phenyl-5-sulfanyl-4H-1,2,4-triazol-3-ol

  • MDL number MFCD00185964
  • Cas Number : 27106-12-3
  • Molecular Weight : 193.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 191-194
Iupac Name 4-phenyl-5-sulfanyl-4H-1,2,4-triazol-3-ol
Inchi Code 1S/C8H7N3OS/c12-7-9-10-8(13)11(7)6-4-2-1-3-5-6/h1-5H,(H,9,12)(H,10,13)
Inchi Key RAOXPTJDYMBTGU-UHFFFAOYSA-N

Description

Melting Point 191-194
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6477574

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6477574
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart