ENAH5802F4E1 Enamine

4-sulfanylpyridine-2,6-dicarboxylic acid

  • MDL number MFCD20646848
  • Cas Number : 871884-79-6
  • Molecular Weight : 199.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-sulfanyl-2,6-pyridinedicarboxylic acid
Inchi Code 1S/C7H5NO4S/c9-6(10)4-1-3(13)2-5(8-4)7(11)12/h1-2H,(H,8,13)(H,9,10)(H,11,12)
Inchi Key ZYCAMEYAPIZEIQ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-216424

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-216424
pictograms GHS05,GHS07
hstatements H302,H312,H315,H318,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart