ENAH5802F4E2 Enamine

2,4-dinitrobenzene-1-thiol

  • MDL number MFCD00797825
  • Cas Number : 2218-96-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-130
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-94691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-94691
Availability details for
Quantity
  • Availability
  • Add to Cart