ENAH5802F4E4 Enamine

4-[3-(dimethylamino)propyl]-5-methyl-4H-1,2,4-triazole-3-thiol dihydrochloride

  • MDL number MFCD26143812
  • Cas Number : 1251923-20-2
  • Molecular Weight : 273.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(3-(dimethylamino)propyl)-5-methyl-4H-1,2,4-triazole-3-thiol dihydrochloride
Inchi Code 1S/C8H16N4S.2ClH/c1-7-9-10-8(13)12(7)6-4-5-11(2)3;;/h4-6H2,1-3H3,(H,10,13);2*1H
Inchi Key OEPAAFYODQDUAN-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-67398

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-67398
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart