ENAH5802F4E7 Enamine

5-[(2-fluorophenyl)methyl]-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD10689213
  • Cas Number : 1092307-62-4
  • Molecular Weight : 210.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66533
Availability details for
Quantity
  • Availability
  • Add to Cart