ENAH5802F4E8 Enamine

5-[(3-fluorophenyl)methyl]-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD10694793
  • Cas Number : 1152542-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-67070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-67070
Availability details for
Quantity
  • Availability
  • Add to Cart