ENAH5802F4EB Enamine

5-(2-cyclohexylethyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD10692292
  • Cas Number : 1152542-39-6
  • Molecular Weight : 212.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66801
Availability details for
Quantity
  • Availability
  • Add to Cart