ENAH5802F4ED Enamine

N,N-dimethyl-4-sulfanylbenzene-1-sulfonamide

  • MDL number MFCD32068937
  • Cas Number : 18325-52-5
  • Molecular Weight : 217.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 100-102
Iupac Name 4-mercapto-N,N-dimethylbenzenesulfonamide
Inchi Code 1S/C8H11NO2S2/c1-9(2)13(10,11)8-5-3-7(12)4-6-8/h3-6,12H,1-2H3
Inchi Key VTIKXMBORKZRBB-UHFFFAOYSA-N

Description

Melting Point 100-102
Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6246143

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6246143
pictograms GHS05,GHS07
hstatements H302,H312,H315,H318,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart