ENAH5802F4EE Enamine

5-(3-phenylpropyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD10695393
  • Cas Number : 1119505-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68263
Availability details for
Quantity
  • Availability
  • Add to Cart