ENAH5802F4EF Enamine

5-(1-phenylpropyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD11201452
  • Cas Number : 1038340-13-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-66758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66758
Availability details for
Quantity
  • Availability
  • Add to Cart