ENAH5802F4F0 Enamine

4-(2-methanesulfonylethyl)-5-methyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD12768058
  • Cas Number : 1179647-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-70563

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70563
Availability details for
Quantity
  • Availability
  • Add to Cart