ENAH5802F4F5 Enamine

3-sulfanyl-4-(2,4,6-trimethylphenyl)-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD17239459
  • Cas Number : 1343723-30-7
  • Molecular Weight : 235.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-mesityl-5-sulfanyl-2,4-dihydro-3H-1,2,4-triazol-3-one
Inchi Code 1S/C11H13N3OS/c1-6-4-7(2)9(8(3)5-6)14-10(15)12-13-11(14)16/h4-5H,1-3H3,(H,12,15)(H,13,16)
Inchi Key YNFHVZUFAZTACU-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7440420

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440420
Availability details for
Quantity
  • Availability
  • Add to Cart