ENAH5802F4F7 Enamine

5-(benzylimino)-4-methyl-4,5-dihydro-1,3,4-thiadiazole-2-thiol

  • Cas Number : 748778-11-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-03479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-03479
Availability details for
Quantity
  • Availability
  • Add to Cart