ENAH5802F4F9 Enamine

5-[(2-chloro-6-fluorophenyl)methyl]-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD10694451
  • Cas Number : 1152542-00-1
  • Molecular Weight : 244.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-66785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-66785
Availability details for
Quantity
  • Availability
  • Add to Cart