ENAH5802F4FC Enamine

tert-butyl 3-(3-sulfanyl-4H-1,2,4-triazol-4-yl)azetidine-1-carboxylate

  • MDL number MFCD31924080
  • Cas Number : 2344679-19-0
  • Molecular Weight : 256.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-(3-mercapto-4H-1,2,4-triazol-4-yl)azetidine-1-carboxylate
Inchi Code 1S/C10H16N4O2S/c1-10(2,3)16-9(15)13-4-7(5-13)14-6-11-12-8(14)17/h6-7H,4-5H2,1-3H3,(H,12,17)
Inchi Key OADWBTFZXDSRGZ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6764520

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6764520
Availability details for
Quantity
  • Availability
  • Add to Cart