ENAH5802F4FE Enamine

tert-butyl 3-(3-sulfanyl-4H-1,2,4-triazol-4-yl)piperidine-1-carboxylate

  • Cas Number : 2344680-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6763629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6763629
Availability details for
Quantity
  • Availability
  • Add to Cart