ENAH5802F500 Enamine

methyl (2S,3R)-3-amino-2-{[(tert-butoxy)carbonyl]amino}butanoate hydrochloride

  • Cas Number : 2343963-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-7427105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7427105
Availability details for
Quantity
  • Availability
  • Add to Cart