ENAH5802F502 Enamine

1-tert-butyl 2-ethyl 3-aminopyrrolidine-1,2-dicarboxylate

  • MDL number MFCD24543791
  • Cas Number : 1786226-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-185650

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185650
Availability details for
Quantity
  • Availability
  • Add to Cart