ENAH581684E3 Enamine

tert-butyl (1R,5S,6R)-6-[(2-chloropyrimidin-4-yl)(methyl)amino]-3-azabicyclo[3.1.0]hexane-3-carboxylate

  • MDL number MFCD32203519
  • Cas Number : 2378490-48-1
  • Molecular Weight : 324.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7446708

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7446708
Availability details for
Quantity
  • Availability
  • Add to Cart