ENAH659198E1 Enamine

2-(2-aminocyclobutyl)acetic acid hydrochloride

  • Molecular Weight : 165.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7466330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7466330
Availability details for
Quantity
  • Availability
  • Add to Cart