ENAH659199C3 Enamine

2-phenyl-3,4-dihydro-2H-1-benzopyran-5-ol

  • MDL number MFCD32219768
  • Cas Number : 92215-50-4
  • Molecular Weight : 226.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7456466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7456466
Availability details for
Quantity
  • Availability
  • Add to Cart