ENAH6591F4CD Enamine

4-bromo-6-fluoro-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD09038174
  • Cas Number : 174603-56-6
  • Molecular Weight : 229.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-3001355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-3001355
Availability details for
Quantity
  • Availability
  • Add to Cart