ENAH6591F5AD Enamine

methyl 4-({[(1-ethyl-1H-pyrazol-4-yl)methyl]amino}methyl)benzoate

  • MDL number MFCD32219795
  • Cas Number : 2402830-76-4
  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7461208

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7461208
Availability details for
Quantity
  • Availability
  • Add to Cart