ENAH6591F5D5 Enamine

tert-butyl 3-(aminomethyl)-3-methylpyrrolidine-1-carboxylate

  • MDL number MFCD12408548
  • Cas Number : 1158758-67-8
  • Molecular Weight : 214.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-1603307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1603307
Availability details for
Quantity
  • Availability
  • Add to Cart