ENAH6591F7EC Enamine

2-(4-fluorophenoxy)benzoic acid

  • MDL number MFCD03840025
  • Cas Number : 2795-63-3
  • Molecular Weight : 232.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-298970

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-298970
Availability details for
Quantity
  • Availability
  • Add to Cart