ENAH6591F973 Enamine

5-chloro-N2-[5-methyl-4-(piperidin-4-yl)-2-(propan-2-yloxy)phenyl]-N4-[2-(propane-2-sulfonyl)phenyl]pyrimidine-2,4-diamine

  • MDL number MFCD26142648
  • Cas Number : 1032900-25-6
  • Molecular Weight : 558.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-180
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7404145

Safety Information

signalword Warning
pcodes P264,P270,P273,P301+P312,P330,P391,P501
MSDS https://www.enaminestore.com/catalog/EN300-7404145
pictograms GHS07,GHS09
hstatements H302,H400,H410
Availability details for
Quantity
  • Availability
  • Add to Cart