ENAH659279D3 Enamine

2-[(tert-butyldimethylsilyl)oxy]-5-methylaniline

  • MDL number MFCD32219711
  • Cas Number : 1841435-42-4
  • Molecular Weight : 237.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7441052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7441052
Availability details for
Quantity
  • Availability
  • Add to Cart