ENAH659288FA Enamine

2-bromo-6-fluoro-4-(trifluoromethyl)benzene-1-sulfonyl chloride

  • MDL number MFCD32199639
  • Cas Number : 2377035-48-6
  • Molecular Weight : 341.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-7440170

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-7440170
Availability details for
Quantity
  • Availability
  • Add to Cart