ENAH9082202B Enamine

(1s,3s)-3-(fluoromethyl)cyclobutan-1-ol

  • Molecular Weight : 104.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1s,3s)-3-(fluoromethyl)cyclobutan-1-ol
Inchi Code 1S/C5H9FO/c6-3-4-1-5(7)2-4/h4-5,7H,1-3H2/t4-,5+
Inchi Key SOVULJRLJUMRNC-SYDPRGILSA-N

Description

Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA

Safety Information

signalword Warning
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P271,P280,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-7562807
hstatements H226,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart